Lead, coarse powder, Supelco®

Danger

1.07362.1000 1.07362.1000
EM1.07362.1000CS 79103.11 USD
CA1.07362.1000 EM1.07362.1000
Lead, coarse powder, Supelco®
Lead

Lead coarse powder GR for analysis.

Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
UN: 3077
ADR: 9,III
Merck Index: 12,05410

Order Now

Specification Test Results

Hill Formula Pb
Molar Mass 207.2 g/mol
Boiling point 1740 °C (1013 hPa)
Density 11.3 g/cm³ (20 °C)
Melting Point 327 °C
Solubility (20 °C) practically insoluble
Storage class 6.1 D Non combustible, acute toxicity cat. 3 / toxic or substances with chronic effects
WGK NWG not water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR