Detector UA™ Urinalysis Reagent Strips, Immunostics
Supplier: Immunostics
IDU-10 IDU-2PG IDU-5OB
10147-128KT 18.91 USD
10147-128 10147-126 10147-130
Detector UA™ Urinalysis Reagent Strips, Immunostics
For the semi-quantitative and qualitative detection of leukocytes, nitrite, urobilinogen, glucose, protein, pH, leukocytes, nitrite, urobilinogen, glucose, protein, pH, blood, specific gravity, ketone, and bilirubin in urine.
Certifications: CLIA Waived. CE Marked.
Ordering information: 100 Urine Reagent Strips, 1 Package Insert, and 1 Dessicant.