Would you like to log in again?
Immuno Lex-ASO™, Immunostics,Inc.
Supplier: IMMUNOSTICS, INC.
A rapid latex agglutination slide test for the qualitative and semi-quantitative detection of Anti-Streptolysin O Antibodies utilizing undiluted samples.
Certifications: CLIA Catergorization: Moderately Complex. CE Marked.
Ordering information: Kit includes 1 Vial of Latex Reagent, 1 Vial of Positive Control, 1 Vial of Negative Control, Disposable Slides, Disposable Plastic Pipettes/Stirrers, and 1 Package Insert. (All Reagent Contain 0.1% Sodium Azide.)
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description No. of Tests Supplier No. VWR Catalog Number Unit Price Quantity
Immuno Lex-ASO™ 50 ASO-50 10147-086 Kit of 50 Retrieving Restricted
Immuno Lex-ASO™ 100 ASO-100 10147-088 Kit of 100 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Sensitivity 200 IU/mL
Time 2 Minutes
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty