Neutralization solution
Synonyms: DNA Neutralizing solutionNeutralization solutionNeutralisation solutionDNA Neutralising solution
351-014-131
10128-430EA 71.61 USD
10128-430
Neutralization solution
Neutralization solution
DNA Neutralizing Solution used to neutralize fractionated DNA contained in an electrophoresis gel after denaturization and prior to transfer to nitrocellulose or nylon membrane
  • Sterile
  • Shelf life 36 months
When utilized in conjunction with a depurination solution, treatment with the denaturing solution results in the hydrolysis of the phosphodiester backbone at the sites of depurination. This allows for a more efficient transfer of large DNA in neutral Southern transfers. Following DNA denaturation, the gel must be neutralized by soaking in DNA Neutralization Solution.
Caution: Research Use Only