ProODO Optical Dissolved Oxygen Handheld Instrument, YSI
626281
10065-234EA
927.28
USD
10065-234
ProODO Optical Dissolved Oxygen Handheld Instrument, YSI
The YSI ProODOTM handheld dissolved oxygen meter provides extreme durability for measuring DO with optical, luminescent based technology for field or lab.
The YSI ProODO dissolved meter with optical oxygen sensor utilizes the ODO® technology in order to take a dissolved oxygen measurement. There is a lot of information available recently about dissolved oxygen technologies among optical, polarographic, and galvanic. Optical DO definitely has advantages in certain applications. We invite you to read on to learn more about the YSI ProODO instrument. The ProODO instrument meets the demands of true field work. The instrument is rated to IP-67 standards even without the battery cover on. In-house testing procedures also include 1-meter drop tests from all angles to ensure expected field-durability.
Other tough features include Optical dissolved oxygen, Military Spec (MS) connectors, Rubber overmolded case, Field tested cables, Rugged, weighted probe guard, 5,000 data-set memory, Expanded DO range of 0-500% (0-50 mg/L), OBOD optical, self-stirring BOD probe for the lab, Password protection, Backlit display and keypad, Graphic display with detailed Help visible with polarized sunglasses, Detailed GLP data, Multiple languages, 100 folders and site lists for logging data with user defined fields, 5,000 data-set memory, Expanded DO range of 0-500% (0-50 mg/L), OBOD optical, self-stirring BOD probe for the lab, Password protection, Backlit display and keypad, Graphic display with detailed Help visible with polarized sunglasses, Detailed GLP data, Multiple languages, and 100 folders and site lists for logging data with user defined fields.
Connect any length cable and probe to the ProODO digital series instrument and use the convenient cable management kit to keep everything simple and organized. Convenient features include cable options from 1- to 100-meters including a cable management kit on 4-meter and longer cables; laboratory BOD probe. Digital based sensors hold calibration data and allow you to easily connect to any ProODO instrument. USB and communicatiosn saddle included. Soft and hard-sided carrying cases. Flow cell. Cable weight. Belt clip. Tripod.
Data Manager desktop software included; 100 user-defined folders and site names Minimum PC requirements; Windows 2000 with SP4 (minimum) or XP with SP2 (minimum); 300 MHz or higher Pentium-compatible CPU; 128 MB of RAM or higher; 80 MB or more of free hard-disk space; USB 2.0; Microsoft.NET (installed with Data Manager).
Probe/Sensor Materials–The following materials may come into contact with the sampling environment: Xenoy® PBT Polycarbonate blend, Noryl, Stainless Steel, Titanium, Polyurethane, Santoprene, Acrylic, Polystyrene. Optional user ID for data security. Built-in barometer.
Certifications: RoHS, CE, WEEE, C-Tick, VCCI, Waterproof IP-67 (even with the battery cover off, floats), 1-meter drop test, Assembled in USA. GLP Compliant.
Accessories information: ProODO Optical DO Field Cables (sold separately); ODO Digital probe assembly with cable. EPA approved for wastewater compliance reporting of DO. The model 626250 cables can be ordered in lengths of 1-, 4-, 10-, 20-, 30-, 40-, 50-, 60-, or 100-meters. Cables come with the ODO optical DO sensor and sensing cap. Cables over 1-meter also come with the model 603062 cable management kit.
Ordering information: When purchasing the ProODO optical DO you will receive the instrument, manual, batteries, communications saddle, USB cable, and Data Manager software. Must order model 626250 cables separately from the ProODO and ProDSS handheld instruments. The model 626250 optical DO cables ONLY work with the ProODO, ProDSS, and 5500D instruments. Contact your VWR representative for more information. Warranty includes 3-year instrument; 2-year cable and probe; 1-year sensor cap.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart