PEEKsil™ Tubing, SGE
Supplier: SGE (Trajan)
0624254 0624245 0624218 0624290 0624378 0624293 0624371 0624304 0624250 0624279 0624352 0624269 0624204 0624354 0624241 0624233 0624311 0624313 0624214 0624227 0624205 0624380 0624315 0624275 0624302 0624232 0624201 0624203 0624351 0624249 0624370 0624226 0624372 0624377 0624312 0624345 0624242 0624217 0624365 0624379 0624273 0624215 0624353 0624366 0624202 0624228 0624216 0624261 0624225 0624350 0624319 0624231 0624252 0624374 0624234 0624342 0624263 0624230 0624367 0624373 0624243 0624301 0624303 0624292 0624376 0624253 0624291 0624262 0624375 0624229 0624265
14233-502PK 86.45 USD
14233-502 14233-622 14233-504 14233-624 14233-506 14233-626 14233-508 14233-628 14233-662 14233-500 14233-620 14226-118 14233-656 14233-612 14233-534 10799-636 14233-614 10799-638 14233-616 14233-618 10799-634 14233-650 14233-610 14233-654 14233-532 14226-108 14226-106 14226-104 14226-102 10799-640 14233-608 10799-642 14226-140 14233-524 10799-648 14233-644 14233-526 14233-646 10799-644 14233-528 14233-604 14233-648 10799-646 14233-606 14233-520 14233-522 14226-138 14226-136 14226-134 14226-132 10799-650 14226-130 10799-652 14233-634 14233-512 10799-658 14233-636 14233-514 14233-638 10799-654 14233-516 10799-656 14233-518 14233-630 14233-632 14233-510 14226-128 14226-124 10799-662 14226-122 10799-660
PEEKsil™ Tubing, SGE
Tubing Chromatography Tubing
PEEKsil™ Tubing is constructed from a core of smooth, precision bore fused silica tubing bonded to a PEEK outer sheath. It is manufactured to exacting tolerances of bore size and concentricity, for reproducible flow rates and reliable connections.
  • Inert smooth flow path, to minimize band broadening
  • Precise inside bore diameter
  • Flexible and robust
  • Lower sample carry-over
  • Color coded for easy identification of internal diameter
  • Perfectly square cut and polished ends
  • Capable of withstanding high pressures
  • Effective pH Range of 0 to 10 (Not compatible with Hydrofluoric acid)
  • Compatible with most organic solvents (unlike PEEK)
The separation achieved is easily lost in tubing and connectors as a result of dispersion. PEEKsil™ Tubing reduces dispersion, therefore reducing the loss of efficiency, sensitivity, and resolution.
Ideal for HPLC and LC-MS applications.
Max. Operating Temperature: 100°C (212°F)
Tolerance: 25μm (Orange): ± 1, 50μm (Beige/Natural): ± 3, 75μm (Black): ± 3, 100μm (Red): ± 3, 150μm (Purple): ± 5, 175μm (Yellow): ± 5, 200μm (Blue): ± 5, 300μm (Grey): ± 5, 530μm (Bone White): ± 10
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: PEEKsil™ Tubing is available in a variety of lengths and internal diameters, allowing for a wide range of system configurations and flow requirements. Tubing int.Ø ranges from 25 to 530 µm, and ext.Ø ranges are available in ¹/₃₂", ¹/₁₆", and 0.36 mm sizes. Overall tubing lengths are available in 50, 100, 150, 250, and 500 mm formats. Custom lengths are also available upon request.