Human Recombinant EGF (from E. coli)
Supplier: Tonbo Biosciences
21-8356-M001 21-8356-U500 21-8356-U100
10208-650EA 98.2 USD
10208-650 10208-652 10002-482
Human Recombinant EGF (from E. coli)
Epidermal growth factor, EGF, is the founding member of the EGF family of proteins. By binding to its receptor epidermal growth factor receptor, EGFR, it stimulates cell growth, proliferation, and differentiation. It is known to be a potent mitogenic factor for cultured cells of ectodermal and mesodermal origin. EGF is involved in wound healing and is known to inhibit gastric secretion.
  • Carrier-free