Human Recombinant IL17F (from E. coli)
Supplier: Tonbo Biosciences
21-8479-U100 21-8479-U025 21-8479-U005 21-8479-U500
10208-296EA 189.83 USD
10208-296 10208-298 10208-300 10002-142
Human Recombinant IL17F (from E. coli)
IL-17F is one of 6 isoforms of the IL-17 family. IL-17F has been shown to be important in inflammatory responses accompanying asthma. IL-17F binds to IL-17RA and IL-17RC receptor isoforms and activates through the MAP kinase pathway. IL-17F is produced by several immune cell lineages including Th17 cells, mast cells, and basophils. Like the entire IL-17 family, IL-17F is implicated in autoimmune inflammatory disease pathogenesis resulting from dysregulated or unmitigated inflammation processes.
  • Carrier-free