Virtuoso™ Vial Identification System, Thermo Scientific
60180-VT100
10061-024EA 13284.9 USD
10061-024
Virtuoso™ Vial Identification System, Thermo Scientific
The Thermo Scientific™ Virtuoso™ Vial Identification System is an automated option to manual vial labeling that turns a necessary chore into a value-added component within a laboratory workflow.
  • Resistant to common laboratory solvents, the vials resist blurring, smudging and withstand the test of time
  • System’s touch screen and network capabilities helps to transfer text, graphics, barcodes, and logos directly onto the vials
  • It is a fast, detailed and reproducible system for producing customised sample information
With advanced labeling capabilities, the Virtuoso system ensures accurate vial identification and reliable data, and increases laboratory productivity.
Ordering information: Convenience Kits include 100 vials and 100 caps. Accessories are available separately.