1,4-Diacryloylpiperazine

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 1,1'-(Piperazine-1,4-diyl)bis(prop-2-en-1-one)

04809237
IC809237EA 217.17 USD
IC809237
1,4-Diacryloylpiperazine
1,4-Diacryloylpiperazine
Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077661
CAS Number: 6342-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR