Coomassie brilliant blue G-250, dark reddish purple powder for electrophoresis C.I. 42655

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Xylene Brilliant Cyanine GC.I. 42655Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium saltSodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonateServa Blue GHydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium saltSodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulfonateAcid Blue 90Brilliant indocyanine GCoomassie Blue G 250Brilliant Blue GBrillant blue G

04808274
IC808274EA 77.41 USD
IC808274
Coomassie brilliant blue G-250, dark reddish purple powder for electrophoresis C.I. 42655
Coomassie brilliant blue G-250

Brilliant blue G is a useful dye for SDS gels as it readily stains proteins with minimal background color. Protein bands can be visualised during staining. It has been used in the Bradford dye-binding protein assay.


A mechanism for dye binding to protein has been proposed, based onmeasuring coomassie brilliant blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids. Brilliant blue G has been used for determining critical micelle concentration of detergents. Recent research reveals CBBG as a P2X7 (P2X purinoceptor 7: P2RX7) antagonist and suggests that it exhibits neuroprotective properties in rat spinal cord cells.


Dark blue solution. Soluble in hot water (50 mg/mL), ethanol (40 mg/mL), methanol and 10% trichloroacetic acid.

Formula: C₄₇H₄₈N₃NaO₇S₂
MW: 854.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078482
CAS Number: 6104-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR