Methylene blue trihydrate, Powder C.I. 52015

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: C.I.52015Swiss BlueBasic blue 9Methylthioninium chloride trihydrate3,7-Bis(dimethylamino)phenothiazin-5-ium chloride trihydrateMethylene blue

04800663
IC800663EA 89.62 USD
IC800663
Methylene blue trihydrate, Powder C.I. 52015
Methylene blue trihydrate

Methylene blue has been used as a stain in bacteriology and as an oxidation-reduction indicator. A sensitive stain for RNA and
ribonuclease. An ethidium bromide alternative for the visualization of nucleic acids in gels:


Gels are viewable in visible light. Does not require a darkroom. No UV induced DNA damage. Gels can be stored for several days. 30 minute destaining time for PAGE gels.


Soluble in water (1 mg/ml)


Caution: Research Use Only (RUO).

Formula: C₁₆H₁₈ClN₃S·3H₂O
MW: 373.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 7220-79-3

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: λmax (water) 662 ± 3 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR