ICNA02960120
0296012001
IC96012001EA 118.97 USD
IC96012001
ICNA02960120
Menhaden fish oil has been refined to produce a more palatable, clearer oil. It has undergone the winterizing process similar to the alkali refined and bleached oil. The cold test results in a minimum of two hours at 40 °F.
Menhaden Oil is rich in omega-3 fatty acids. When used to partially supplement a high-fat diet in diabetic rats. Menhaden oil improved end points for peripheral neuropathy by improving motor and sensory nerve conduction velocity. Decreased accumulation of fatty acids in rat livers was also demonstrated.
Menhaden oil is use as a control or check sample for fatty acid methyl ester (FAME) analyses.
Caution: Store at -20 °C, store under nitrogen.