Vanderzant modification vitamin mixture for insects, MP Biomedeicals
0290324401 0290324405
IC90324401EA 272.06 USD
IC90324401 IC90324405
Vanderzant modification vitamin mixture for insects, MP Biomedeicals
Vanderzant Vitamin Mixture for insects is an animal-derived component free vitamin mixture developed or use in insect cell culture.
  • Store at Room Temperature (15-30 °C)
  • Powder
It is a mixture of 12 vitamins needed for insect cell culture. For full description see product datasheet.