Corn (Ground, Yellow), granules, MP Biomedicals

0290141180 0290141125
IC90141180EA 194.54 USD
IC90141180 IC90141125
Corn (Ground, Yellow), granules, MP Biomedicals
Corn ( Ground, yellow) is used as pet nutrition and natural pet food.

  • Store at room temperature (15-30 °C)
  • Yellow and White Granules

Components: Carbohydrate 77.1%; Protein 8.0%; Fat 2.3%; Fiber 0.8%; Water 11.1%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR