Alphacel Non-Nutritive Bulk
0290045325 0290045380 0290045305
IC90045325EA 170.5 USD
IC90045325 IC90045380 IC90045305
Alphacel Non-Nutritive Bulk
An extremely fine, white, odorless tasteless, water insoluble, chemically pure cellulose powder, manufactured from natural fibers.
  • Presentation: White Powder
  • Store at room temperature (15-30 ºC)
Alphacel Non-Nutritive Bulk is used to study the effect of conjugated linoleic acids from beef or industrial hydrogenation on growth and adipose tissue characteristics of rats. It is also used as a harmless, biologically acceptable, bulking agent in the specialized diets of laboratory animals.
Alphacel is manufactured by a dry, mechanical process from bleached and purified wood pulp.