Sodium dihydrogen phosphate, anhydrous ≥98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium dihydrogen phosphateSodium phosphate monobasicSodium dihydrogen orthophosphateMonobasic Sodium PhosphateSodium dihydrogenphosphate (granular)

0219550090 0219550091 0219550092 0219550080
IC19550090EA 130.94 USD
IC19550090 IC19550092 IC19550091 IC19550080
Sodium dihydrogen phosphate, anhydrous ≥98%, white powder
Sodium dihydrogen phosphate

Sodium phosphate monobasic is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography.


Useful in conjunction with Sodium Dibasic Phosphate, in the preparation of biological buffers. Used in many applications including the purification of antibodies.


Freely soluble in water; insoluble in ethanol.


Caution: Research Use Only (RUO).

Formula: NaH₂PO₄
MW: 119.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
https://us.vwr.com/stibo/web/std.lang.all/66/33/36426633.jpg

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
Moisture Content ≤1%
pH (1% aq soln) 3 - 5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR