DEAE-Dextran chloride form, off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Diethylaminoethyl dextran

0219513380 0219513350 0219513310
IC19513380EA 411.25 USD
IC19513380 IC19513350 IC19513310
DEAE-Dextran chloride form, off-white powder
DEAE-Dextran

DEAE-Dextran, a polycationic diethylaminoethyl ether of dextran is a stabilizing molecule and transfection reagent. The diethylaminoethyl groups are linked to glucose residues by ether linkages.


Prepared from dextran MW: 500 000
Nitrogen: ~ 3,2%


DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. The use of DEAE-Dextran as an efficient adjuvant in vaccine production is well established. For instance, DEAE-Dextran has been used for immunization of guinea pigs and swine with inactivated foot-and-mouth disease and rhesus monkeys with inactivated Venezulean equine encephalomyelitis virus, IVEE. The mechanism of action of DEAE-Dextran is not clear. The effect on the humoral response of rhesus monkeys to IVEE vaccine results in a typical IgM-IgG response. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.


Soluble in water (100 mg/ml) and dilute salt solutions. Solutions can be sterilized by autoclaving even at low pH. Autoclaving can be done at 110 to 115 °C. For 20% (w/v) or less solutions autoclave for approximately 30 minutes; for solutions above 20%, autoclave for approximately 15 minutes. Cool slowly.

Order Now

Specification Test Results

Identity Test Passes
Nitrogen 3 - 4%
pH 5.0 - 7.0
Loss on Drying ≤10%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR