Coenzyme Q10 ≥99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (all-E)-2-(3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dioneUbidecarenoneUbiquinone 10

0219510825
IC19510825EA 70.35 USD
IC19510825
Coenzyme Q10 ≥99%, powder
Coenzyme Q10

Soluble in chloroform (50 mg/mL - clear to slightly hazy, yellow to orange solution), ethanol or lipids; practically insoluble in water.


Arsenic (As2O3) 2 ppm

Formula: C₅₉H₉₀O₄
MW: 863.37 g/mol
MDL Number: MFCD00042919
CAS Number: 303-98-0

Order Now

Specification Test Results

Synonyms Ubiquinone 50; Q-10; Ubiquinone 10; Ubidecarenone
Form Orange Powder
Storage Store at -0 °C
Extinction Coefficient E1% ≥162 at 275 nm in EtOH (Lit.)
Heavy Metals Heavy Metals (as Pb) 10 ppm
Moisture Content 0.1%
Residue on Ignition 0.2%
Source Bovine Heart

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR