Isodesmosine ≥99%, yellow solid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-(4-Amino-4-carboxybutyl)-1-(5-amino-5-carboxypentyl)-3,5-bis(3-amino-3-carboxypropyl)pyridinium chloride2-amino-6-[3,5-bis(3-amino-4-hydroxy-4-oxobutyl)-2-(4-amino-5-hydroxy-5-oxopentyl)pyridin-1-ium-1-yl]hexanoic acidIso-desmosin

0219137901 0219137910 0219137905
IC19137905EA 2035.8 USD
IC19137905 IC19137910 IC19137901
Isodesmosine ≥99%, yellow solid
Isodesmosine

Isodesmosine is prepared according to the method of Starcher, et al. from hydrolysed bovine neck ligament by solvent extraction and chromatography on Dowex 50. It is chromatographically purified hygroscopic crystalline powder.


Caution: Research Use Only (RUO).

Formula: C₂₄H₄₀N₅O₈+
MW: 526.611 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00211039
CAS Number: 991-01-5

Order Now

Specification Test Results

Identity Test Passes
Source Bovine Neck Ligament
Purity (amino acid analysis) ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR