Serum, Goat is suitable replacement for FBS on most cell lines and primary cultures for Biomedical Research - virus isolation and identification, virological studies, etc
- Presentation: Frozen Red Liquid
- pH: 6.5 - 8.0
It is used as an effective NBCS replacement for the culture of fish cells and could be successfully used in place of fetal bovine serum in the growth medium for long-term in vitro propagation of T. annulata schizontsThis is used for blocking agents and controls in immunoassays.
Osmolality-275 - 340 mOsm/kg , Protein- 5.5 - 8.0 g/dl , Hemoglobin- 20 mg/dl
Goat Serum is a pooled preparation obtained from a normal donor herd.