Acriflavine hydrochloride, redish brown powder

Danger

Synonyms: 3,6-Diamino-10-methylacridinium chloride hydrochlorideEuflavine

0219067525
IC19067525EA 70.99 USD
IC19067525
Acriflavine hydrochloride, redish brown powder
Acriflavine hydrochloride

Acriflavine Hydrochloride is a mixture of 3,6-diamino-10-methylacridinium chloride and 3,6-diaminoacridine (approximately an 9 to 1 mixture respectively).


  • UV/Visible Absorbance: λ max (meOH) 460 ± 4 nm
  • Soluble in water (1 mg/mL- clear, dark orange solution).


Acriflavine Hydrochloride acts as an intercalating dye and also reported to inhibit mitochondriogenesis. It is an antiseptic and has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.

Formula: C₂₇H₂₈Cl₄N₆
MW: 578.36 g/mol
Melting Pt: 260 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00069039
CAS Number: 8063-24-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: l max (meOH) 460±4nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR