Retinol all trans all trans (synthetic), orange powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
(2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol
, all-trans-Retinol
, Vitamin A alcohol
Retinol is one of the animal forms of vitamin A. It is a diterpenoid and an alcohol.
Retinol is an effective antioxidant displaying lipoperoxy radical scavenging activity. The interactions between Retinol and Vitamin E (α-tocopherol) in suppressing lipid peroxidation were observed in bovine retinal membrane preparations. It may influence the production of transition vesicles by stimulating the activity of a protein disulfide isomeraselike activity involved in vesicle formation. Retinol (10 µM) stimulated DNA synthesis and possibly repair mechanisms in Sertoli cells of rat.
Biochem/physiol Actions: Ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR) that act as transcription factors to regulate the growth and differentiation of normal and malignant cells.
Soluble in chloroform (50 mg/ml), absolute alcohol, methanol, ether, fats and oils; practically insoluble in water and glycerol.
Formula:
C₂₀H₃₀O MW: 286.46 g/mol Melting Pt: 61…63 °C (lit.) Storage Temperature: Freezer |
MDL Number:
MFCD00001552 CAS Number: 68-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...