ICNA02190213
0219021325
IC19021325EA 358.75 USD
IC19021325
ICNA02190213
Soluble in alcohol, acetone, ether, chloroform, propylene glycol, polyethylene glyrols, olive oil, cotton seed oil, dil aq sol in alkalies, methanol (50 mg/ ml-clear, light yellow solution); practically insoluble in water