Lidocaine, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0219011183
0219011125
IC19011183EA
427.7
USD
IC19011183
IC19011125
Lidocaine, white powder
Lidocaine
Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.
Lidocaine is used to treat allergies. It is generally used to differentiate and identify leucocytes, malaria parasites, and trypanosomas.
Soluble to 100 mM in ethanol.
Caution: Research Use Only (RUO).
Formula:
C₁₄H₂₂N₂O Melting Pt: 68.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026733 CAS Number: 137-58-6 |
Specification Test Results
Identity Test | Passes |
Identity Test | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...