ICNA02190052
0219005205 0219005225
IC19005205EA 66.01 USD
IC19005205 IC19005225
ICNA02190052
α-Cyclodextrin is reported useful for the selective precipitation of enantiomeric, positional or structural isomers. Cyclodextrins have hydrophobic cavities; form inclusion compounds with organic substances, salts and halogens in the solid state or in aqueous solutions.
Soluble in water (50 mg/mL).