Kifunensine ≥98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215995201
IC15995201EA 277.31 USD
IC15995201
Kifunensine ≥98%, white powder
Kifunensine

Kifunensine is an alkaloid compound, originally isolated from the actinomycete Kitasporia kifunensis, that exhibits potent and selective inhibition of class I α-mannosidases. Kifunensine is used to suppress Endoplasmic Reticulum-Associated Degradation (ERAD) via the inhibition of endoplasmic reticulum-associated mannosidase activity.


Selective inhibitor of class I glycoprotein-processing α-mannosidases


Soluble in water (25 mg/ml).

Formula: C₈H₁₂N₂O₆
Storage Temperature: Freezer
CAS Number: 109944-15-2
https://us.vwr.com/stibo/web/std.lang.all/97/31/36419731.jpg

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR