Victoria blue B, dark brown powder C.I. 44045

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215829125
IC15829125EA 56.6 USD
IC15829125
Victoria blue B, dark brown powder C.I. 44045
Victoria Blue B

Victoria Blue B is used as a nuclear stain in cytology/histochemical applications. Victoria Blue B may be used in the quantitative determination of phospholipids.


More soluble in the lower aliphatic alcohols; slightly soluble in water.


Caution: Research Use Only (RUO).

Formula: C₃₃H₃₂ClN₃
Storage Temperature: Ambient
MDL Number: MFCD00011878
CAS Number: 2580-56-5

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: λmax (ethanol) Report Resut

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR