Iron(III) sulfate hydrate, pale gray powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Ferric sulfate hydrate
, Diiron trissulfate hydrate
, Diiron trissulphate hydrate
, Ferric sulphate hydrate
Ferric Sulfate is used in dyeing as a mordant, and as a coagulant for industrial wastes. It is also used in pigments, and in pickling baths for aluminum and steel. Medically it is used as an astringent and styptic. It also acts as an important oxidative agent.
Soluble in water; sparingly soluble in alcohol; practically insoluble in acetone, ethyl acetate.
Formula:
Fe₂(SO₄)₃·xH₂O MW: 399.88 g/mol (anhy) Melting Pt: 1539 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00149714 CAS Number: 15244-10-7 |
Specification Test Results
Identity Test | Passes |
Moisture | Record Result |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...