Ethoxyquin ≥90%, viscous liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline
, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline
, 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline
, Ethoxyquin
0215796380
IC15796380EA
121.59
USD
IC15796380
Ethoxyquin ≥90%, viscous liquid
Ethoxyquin
Ethoxyquin is a quinoline-based antioxidant.
Ethoxyquin is an antioxidant which inhibits chemically induced carcinogenesis, where it exerts its chemopreventive action by inducing glutathione synthetase and the phase II drug metabolizing enzyme, glutathione S-transferase (GST) and an antidegradation agent for rubber.
Formula:
C₁₄H₁₉NO |
MDL Number:
MFCD00023883 CAS Number: 91-53-2 |
Specification Test Results
Identity Test | Passes |
Purity | ≥90% |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...