ICNA02157755
0215775580
IC15775580EA 368.7 USD
IC15775580
ICNA02157755
N,N`-Diisopropylcarbodiimide is used as an alternative to dicyclohexylcarbodiimide in peptide synthesis. Also as coupling reagent for peptide syntheses.