Glycerol-3-Phosphate Oxidase
Supplier: MP Biomedicals
0215724080 0215724050 0215724090
IC15724050EA 57.84 USD
IC15724050 IC15724080 IC15724090
Glycerol-3-Phosphate Oxidase
Soluble (0.45 unit/mL) in cold 20 mM Tris HCl, pH 7.5 at 37 °C, containing 0.2% (w/v) bovine serum albumin.
One unit will oxidize 1.0 µmole of L-glycerol 3-phosphate to dihydroxyacetone phosphate with the formation of H2O2 per min at 37 °C.
Glycerol-3-phosphate oxidase catalyzes the conversion of sn-glycerol-3-phosphate to glycerone phosphate and peroxide.