Sudan orange G, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215670425
IC15670425EA 102.77 USD
IC15670425
Sudan orange G, Powder
Sudan Orange G
Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
MDL Number: MFCD00002275
CAS Number: 2051-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR