Probenecid, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 4-[(dipropylamino)sulphonyl]benzoic acid

0215637080 0215637025 0215637090
IC15637080EA 348.6 USD
IC15637080 IC15637090 IC15637025
Probenecid, white powder
Probenecid

Probenecid is useful as an inhibitor of the organic anion transporter.


Probenecid is an inhibitor of multidrug resistance-associated proteins (Mdr-1). This compound blocks the active site of Mdr-1 and prevents cellular efflux of other active compounds, enhancing their effectiveness by allowing them to remain in the cytoplasm.


Soluble in dilute alkalies; slightly soluble ether and acetone; sparingly soluble in chloroform and ethanol; practically insoluble in water and dilute acids.

Formula: C₁₃H₁₉NO₄S
MW: 285.4 g/mol
Melting Pt: 381…385 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00038402
CAS Number: 57-66-9
https://us.vwr.com/stibo/web/std.lang.all/99/93/36419993.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR