Potassium hydrogen sulfate, crystals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Potassium bisulphatePotassium bisulfate

0215634083
IC15634083EA 123.26 USD
IC15634083
Potassium hydrogen sulfate, crystals
Potassium hydrogen sulphate

Soluble in Water (100 mg/mL, 36%).


acid number: 35‑37%(as H2SO4)(Lit.)

Formula: KHSO₄
MW: 136.17 g/mol
MDL Number: MFCD00011404
CAS Number: 7646-93-7
UN: 2509
ADR: 8,II

Order Now

Specification Test Results

Form Clear Crystals
Storage Store at Room Temperature (15-30 °C).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR