N,N-Diethyl-4-nitrosoaniline, powder

Synonyms: 4-Nitroso-N,N-diethylaniline

0215592750
IC15592750EA 181.73 USD
IC15592750
N,N-Diethyl-4-nitrosoaniline, powder
N,N-Diethyl-4-nitrosoaniline

(N,N-Diethyl-4-nitrosoaniline)

Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
MDL Number: MFCD00002064
CAS Number: 120-22-9
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR