3-Mercapto-1,2-propanediol ≥98%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (±)-3-Mercapto-1,2-dihydroxy propane(±)-3-Mercapto-1,2-propanediol(±)-α-Monothioglycerinealpha-Thioglycerolα-Monothioglycerol(±)-1-Thioglycerine1-ThioglycerolMonothioglycerol

0215572380 0215572325 0215572350 0215572383
IC15572350EA 111.29 USD
IC15572350 IC15572383 IC15572380 IC15572325
3-Mercapto-1,2-propanediol ≥98%, liquid
3-Mercapto-1,2-propanediol

α-Monothioglycerol is a reagent that is used in analytical chemistry, material science, and cell culture research.

Formula: C₃H₈O₂S
MW: 108.16 g/mol
MDL Number: MFCD00004879
CAS Number: 96-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR