ICNA02155154
0215515480
IC15515480EA 1087.87 USD
IC15515480
ICNA02155154
Ketoprofen is a non-selective NSAID which inhibits both human recombinant Cox-1 (IC50 of 0.5 μM) and Cox-2 (IC50 of 2.33 μM).
Ketoprofen is used for its antipyretic, analgesic, and anti-inflammatory properties by inhibiting Cox-1 and Cox-2 (cyclooxygenase-1 and -2) enzymes reversibly, which decreases production of pro-inflammatory prostaglandin precursors.
Non-steroidal anti-inflammatory compound that is selective for COX-1. It acts by inhibiting the body's production of prostaglandin.
Soluble in ethanol (50 mg/ml), acetone.(Lit.)