Chrysin, yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 5,7-Dihydroxyflavone5,7-Dihydroxy-2-phenyl-4-benzopyrone5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one5,7-Dihydroxy-2-phenyl-4H-chromen-4-one5,7-Dihydroxy-2-phenylchromen-4-one

0215498525 0215498505
IC15498505EA 48.55 USD
IC15498505 IC15498525
Chrysin, yellow powder
Chrysin
Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
Melting Pt: 284…290 °C (lit.)
MDL Number: MFCD00006834
CAS Number: 480-40-0
EINECS: 207-549-7

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR