ε-Caprolactam, solid

Synonyms: Azepan-2-one

0215492683
IC15492683EA 33.27 USD
IC15492683
ε-Caprolactam, solid
ε-Caprolactam

Soluble in water (50 mg/mL).

Formula: C₆H₁₁NO
MW: 113.16 g/mol
MDL Number: MFCD00006936
CAS Number: 105-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR