2-Bromethylamine hydrobromide ≥98.0%, white crystalline powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
2-Bromoethylamine hydrobromide
, 2-Bromoethylammonium bromide
0215487525
IC15487525EA
42.46
USD
IC15487525
2-Bromethylamine hydrobromide ≥98.0%, white crystalline powder
2-Bromethylamine hydrobromide
2-Bromoethylamine is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone.
Caution: Research Use Only (RUO).
Formula:
BrCH₂CH₂NH₃Br MW: 204.89 g/mol Melting Pt: 342…345° F (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00012886 CAS Number: 2576-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...