Roseoflavin ≥97%, brown powder

0215425280
IC15425280EA 2307.5 USD
IC15425280
Roseoflavin ≥97%, brown powder
Roseoflavin

Roseoflavin is a weak antibacterial agent active against some gram-positive bacteria, especially Staphylococcus aureus, Sarcina latea, Bacillus cereus and Bacillus subtilis. Also exhibits antiriboflavin activity and inhibits the biosynthesis of riboflavin by a repressor mechanism.


Caution: Research Use Only (RUO). Protect from light.

Formula: C₁₈H₂₃N₅O₆
Storage Temperature: Refrigerator
CAS Number: 51093-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR