1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, Powder
Synonyms:
2,3,5-Tri-o-Benzoyl-1-o-Acetyl-β-D-Ribofuranose
, β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate
, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose
, 1-O-Acetyl-2,3,5-Tri-O-benzoyl-beta-D-Ribofuranose
0215395525
IC15395525EA
296.39
USD
IC15395525
1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, Powder
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose
Starting material for nucleoside synthesis.
Formula:
C₂₈H₂₄O₉ MW: 504.5 g/mol |
MDL Number:
MFCD00005357 CAS Number: 6974-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...