3,4-Dihydroxybenzylamine hydrobromide, Powder

Synonyms: 4-(Aminomethyl)benzene-1,2-diol hydrobromide

0215374025
IC15374025EA 38.52 USD
IC15374025
3,4-Dihydroxybenzylamine hydrobromide, Powder
3,4-Dihydroxybenzylamine hydrobromide

Soluble in water (50 mg/mL) or ethanol. Dissolve in oxygen-free water containing 0.1% sodium metabisulfite or other antioxidants. Solutions should be freshly prepared and protected from exposure to light.

Formula: C₇H₁₀BrNO₂
MW: 220.07 g/mol
MDL Number: MFCD00012859
CAS Number: 16290-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR