3-(4-Fluorobenzoyl)propionic acid, solid

Synonyms: R 11302Haloperidol Metabolite III

0215369310
IC15369310EA 46.33 USD
IC15369310
3-(4-Fluorobenzoyl)propionic acid, solid
3-(4-Fluorobenzoyl)propionic acid

Applications: Administration of haloperidol Metabolite III effected a suppression of locomotor activity and induced catalepsy in a manner similar to that observed with haloperidol.
Soluble in methanol or dilute aqueous base; insoluble in water.

Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
MDL Number: MFCD00002793
CAS Number: 366-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR