Glucagon

Supplier: MP Biomedicals
0215287901
IC15287901EA 1069.29 USD
IC15287901
Glucagon
  • Antihypoglycemic agent
  • Amino Acid Sequence: His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr
  • Soluble in 1 % acetic acid (1 mg/mL-clear colorless solution)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR