Methyl blue, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Disodium [[4-[bis[4-[(sulphonatophenyl)amino]phenyl]methylene]cyclohexa-2,5-dien-1-ylidene]amino]benzenesulphonatePoirriers blueAniline blue sodium saltCotton blueWater blueC.I. 42780Aniline blue, water solubleAcid Blue 93

0215264325
IC15264325EA 115.82 USD
IC15264325
Methyl blue, powder
Methyl blue

Soluble in water (1 mg/ 1 mL).

Formula: C₃₇H₂₇N₃Na₂O₉S₃
MW: 799.81 g/mol
MDL Number: MFCD00058509
CAS Number: 28983-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR