ICNA02152641
0215264125
IC15264125EA 107.42 USD
IC15264125
ICNA02152641
Acid Green 5 is useful as a general plasma stain, as a histological stain, and a cytological counterstain. Can be use as a cytological counterstain and as a substitute for aniline blue in Gomori′s one-step trichrome procedure.
Soluble in water.
Caution: Research Use Only (RUO).