Basic blue 3, dark green powder C.I. 51004

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: BasicblueBGCI BASIC BLUE 3BLUE DE LYON

0215262910
IC15262910EA 43.51 USD
IC15262910
Basic blue 3, dark green powder C.I. 51004
Basic blue 3

Soluble in water, EGME, ethanol.


Caution: Research Use Only (RUO).

Formula: C₂₀H₂₆N₃OCl
MW: 359.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011928
CAS Number: 33203-82-6

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: λmax (water) Report Result

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR