Alcian blue 8GX, dark blue powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
C.I. 74240
, C.N. AnAn-6
, Ingrain Blue 1
, Alcian blue 8 GX
Alcian Blue 8GX is a cationic dye used to quantitatively determine glycosaminoglycans by forming insoluble complexes in solution.
Alcian Blue 8GX can be used to determine proteins based on their Resonance Light Scattering by strongly enhancing the signals. It is useful as a bacterial stain, and as a dye for histiocytes and fibroblasts. Alcian Blue 8GX is used as a heteroglycan stain for neutral, sulfated and phosphated mucopolysaccharides and glycosaminoglycans in tissues such as cartilage and extracellular matrices.
Soluble in absolute ethanol, cellosolve, ethylene glycol, water (8 mg/ml) or ethanol (1 mg/ml); practically insoluble in xylene
Caution: Research Use Only (RUO).
Formula:
C₅₆H₆₈Cl₄CuN₁₆S₄ MW: 1298.86 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010720 CAS Number: 33864-99-2 |
Specification Test Results
Identity Test | Passes |
UV/VIS: λmax (water) | 620 - 630 nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...